siRNA Details
| Gene Symbol: NCAPG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281932.1 | NP_001268861.1 | 6 | 723 | ||
| NM_001281933.1 | NP_001268862.1 | 5 | 670 | |||
| NM_017760.6 | NP_060230.5 | 5 | 670 | |||
| NR_104054.1 | 7 | 824 | ||||
| XM_005249547.2 | XP_005249604.1 | 5 | 668 | |||
| XM_011516356.2 | XP_011514658.1 | 5 | 668 | |||
| XM_011516357.2 | XP_011514659.1 | 6 | 696 | |||
| XM_011516358.2 | XP_011514660.1 | 5 | 668 | |||
| XM_017012374.1 | XP_016867863.1 | 5 | 668 | |||
| XM_017012375.1 | XP_016867864.1 | 5 | 668 | |||
| XM_017012376.1 | XP_016867865.1 | 5 | 668 | |||
| XM_017012378.1 | XP_016867867.1 | 3 | 287 | |||
| GenBank mRNA | AK092008.1 | - | 6 | 658 | ||
| AK315699.1 | - | 3 | 287 | |||
| BC043404.1 | - | 5 | 611 | 6045248 | ||
| BX537845.1 | - | 5 | 637 | |||

NCAPG2
Genomic Map