siRNA Details
| Gene Symbol: CDKAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017774.3 | NP_060244.2 | 5 | 527 | ||
| XM_011514718.1 | XP_011513020.1 | 5 | 527 | |||
| XM_011514719.2 | XP_011513021.1 | 5 | 527 | |||
| XM_017010986.1 | XP_016866475.1 | 5 | 527 | |||
| XM_017010987.1 | XP_016866476.1 | 5 | 524 | |||
| GenBank mRNA | AK000349.1 | - | 5 | 465 | ||
| AK128546.1 | - | 4 | 414 | |||
| AK291735.1 | - | 5 | 507 | |||
| BC121020.1 | - | 5 | 457 | 40117357 | ||
| BC121021.1 | - | 5 | 457 | 40117363 | ||

CDKAL1
Genomic Map