siRNA Details
| Gene Symbol: TTC19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271420.1 | NP_001258349.1 | 10 | 1707 | ||
| NM_017775.3 | NP_060245.3 | 10 | 1570 | |||
| GenBank mRNA | AK000350.1 | - | 8 | 786 | ||
| AK055780.1 | - | 10 | 1263 | |||
| AK056878.1 | - | 10 | 1482 | |||
| AK094819.1 | - | 3 | 1764 | |||
| AK297783.1 | - | 10 | 1126 | |||
| AL832412.1 | - | 2 | 1495 | |||
| BC011698.2 | - | 10 | 1109 | |||
| BC073796.1 | - | 10 | 1113 | |||
| BC105128.1 | - | 10 | 1570 | 8144131 | ||
| BC112107.1 | - | 10 | 1570 | 8327575 | ||

TTC19
Genomic Map