siRNA Details
| Gene Symbol: SPDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017785.4 | NP_060255.3 | 9 | 1360 | ||
| XM_011534585.1 | XP_011532887.1 | 9 | 1306 | |||
| XM_011534586.2 | XP_011532888.1 | 9 | 1242 | |||
| XM_011534587.2 | XP_011532889.1 | 10 | 1507 | |||
| XM_011534588.2 | XP_011532890.1 | 10 | 1445 | |||
| XM_011534589.2 | XP_011532891.1 | 9 | 1255 | |||
| XM_011534590.2 | XP_011532892.1 | 8 | 996 | |||
| XM_017009601.1 | XP_016865090.1 | 9 | 1191 | |||
| XM_017009602.1 | XP_016865091.1 | 8 | 996 | |||
| GenBank mRNA | AF269167.1 | - | 7 | 954 | ||
| AK000371.1 | - | 8 | 1086 | |||
| AK074227.1 | - | 6 | 765 | |||
| AK304623.1 | - | 8 | 1024 | |||
| AK310442.1 | - | 9 | 1159 | |||
| AY517557.1 | - | 9 | 1191 | |||
| BC012568.1 | - | 9 | 1226 | 4102558 | ||
| BU166207.1 | - | 3 | 301 | |||

SPDL1
Genomic Map