siRNA Details
Gene Symbol: RPP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017793.2 | NP_060263.2 | 1 | 2107 | ||
GenBank mRNA | AY034074.1 | - | 2 | 1161 | ||
BC002497.2 | - | 1 | 1425 | 3050784 | ||
BC007270.1 | - | 1 | 1437 | 3160319 | ||
BE254727.1 | - | 1 | 362 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
