siRNA Details
| Gene Symbol: YTHDF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017798.3 | NP_060268.2 | 4 | 1239 | ||
| GenBank mRNA | AB055518.1 | - | 1 | 580 | ||
| AK000398.1 | - | 2 | 399 | |||
| AK095795.1 | - | 4 | 957 | |||
| AK300073.1 | - | 2 | 969 | |||
| AL834366.1 | - | 1 | 360 | |||
| BC003681.1 | - | 3 | 913 | |||
| BC016920.2 | - | 1 | 656 | |||
| BC025264.1 | - | 1 | 776 | |||
| BC050284.1 | - | 4 | 1239 | 5298438 | ||

YTHDF1
Genomic Map