siRNA Details
| Gene Symbol: TMEM161A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256766.1 | NP_001243695.1 | 5 | 377 | ||
| NM_017814.2 | NP_060284.1 | 7 | 686 | |||
| XM_011528117.2 | XP_011526419.1 | 2 | 447 | |||
| XM_017026916.1 | XP_016882405.1 | 7 | 582 | |||
| GenBank mRNA | AK000429.1 | - | 7 | 643 | ||
| AK096964.1 | - | 5 | 342 | |||
| AY358088.1 | - | 7 | 670 | |||
| BC005210.1 | - | 7 | 646 | 4041226 | ||
| BC016478.2 | - | 6 | 610 | |||

TMEM161A
Genomic Map