siRNA Details
| Gene Symbol: TMEM161A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256766.1 | NP_001243695.1 | 9 | 868 | ||
| NM_017814.2 | NP_060284.1 | 11 | 1177 | |||
| XM_011528117.2 | XP_011526419.1 | 6 | 938 | |||
| XM_017026916.1 | XP_016882405.1 | 11 | 1073 | |||
| GenBank mRNA | AK000429.1 | - | 11 | 1134 | ||
| AK096964.1 | - | 9 | 833 | |||
| AK225424.1 | - | 11 | 1091 | |||
| AY358088.1 | - | 11 | 1161 | |||
| BC005210.1 | - | 11 | 1137 | 4041226 | ||
| BC016478.2 | - | 10 | 1101 | |||

TMEM161A
Genomic Map