siRNA Details
| Gene Symbol: RHBDL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304746.1 | NP_001291675.1 | 8 | 1078 | ||
| NM_017821.4 | NP_060291.2 | 8 | 1108 | |||
| GenBank mRNA | AK097621.1 | - | 8 | 1843 | ||
| AK294115.1 | - | 8 | 1078 | |||
| AY126343.1 | - | 7 | 834 | |||
| BC137108.1 | - | 7 | 957 | 9021105 | ||
| BC137110.1 | - | 7 | 957 | 9021107 | ||
| BC144476.1 | - | 7 | 944 | |||

RHBDL2
Genomic Map