siRNA Details
| Gene Symbol: PARP16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316943.1 | NP_001303872.1 | 2 | 700 | ||
| NM_017851.5 | NP_060321.3 | 2 | 700 | |||
| XM_006720589.2 | XP_006720652.1 | 2 | 741 | |||
| XM_006720590.2 | XP_006720653.1 | 2 | 741 | |||
| XM_017022388.1 | XP_016877877.1 | 2 | 741 | |||
| XM_017022389.1 | XP_016877878.1 | 2 | 741 | |||
| GenBank mRNA | AK058010.1 | - | 2 | 284 | ||
| BC006389.1 | - | 2 | 546 | 4127509 | ||
| BC031074.1 | - | 2 | 349 | 5274010 | ||

PARP16
Genomic Map