siRNA Details
| Gene Symbol: ODAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017855.3 | NP_060325.3 | 2, 3 | 116 | ||
| XM_005265698.3 | XP_005265755.1 | 3, 4 | 180 | |||
| XM_006714245.3 | XP_006714308.1 | 3, 4 | 157 | |||
| GenBank mRNA | BG194365.1 | - | 2, 3 | 119 | ||
| EF113908.1 | - | 2, 3 | 118 | |||

ODAM
Genomic Map