siRNA Details
| Gene Symbol: ODAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017855.3 | NP_060325.3 | 5, 6 | 443 | ||
| XM_005265698.3 | XP_005265755.1 | 6, 7 | 507 | |||
| XM_006714245.3 | XP_006714308.1 | 6, 7 | 484 | |||
| XM_017008331.1 | XP_016863820.1 | 4, 5 | 399 | |||
| GenBank mRNA | AK000520.1 | - | 2, 3 | 94 | ||
| AK225547.1 | - | 2, 3 | 74 | |||
| BC017796.1 | - | 1, 2 | 50 | 4689080 | ||
| BG194365.1 | - | 5, 6 | 446 | |||
| EF113908.1 | - | 5, 6 | 445 | |||

ODAM
Genomic Map