siRNA Details
| Gene Symbol: TTC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318533.1 | NP_001305462.1 | 13 | 1252 | ||
| NM_017868.3 | NP_060338.3 | 13 | 1234 | |||
| XM_005271604.3 | XP_005271661.1 | 13 | 1450 | |||
| XM_011542890.2 | XP_011541192.1 | 14 | 1448 | |||
| XM_017017943.1 | XP_016873432.1 | 13 | 1234 | |||
| XM_017017944.1 | XP_016873433.1 | 14 | 1804 | |||
| XM_017017945.1 | XP_016873434.1 | 14 | 1287 | |||
| XM_017017946.1 | XP_016873435.1 | 13 | 1457 | |||
| XM_017017947.1 | XP_016873436.1 | 13 | 1234 | |||
| XM_017017948.1 | XP_016873437.1 | 13 | 1234 | |||
| XM_017017949.1 | XP_016873438.1 | 13 | 1234 | |||
| XM_017017950.1 | XP_016873439.1 | 13 | 1234 | |||
| GenBank mRNA | AK000542.1 | - | 12 | 1126 | ||
| AK023921.1 | - | 13 | 1180 | |||
| AK091887.1 | - | 15 | 1846 | |||
| AK223117.1 | - | 12 | 1126 | |||
| AK292331.1 | - | 13 | 1172 | |||
| BC011546.1 | - | 12 | 1176 | |||
| BC032355.1 | - | 12 | 1156 | 5219117 | ||

TTC12
Genomic Map