siRNA Details
| Gene Symbol: CLN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017882.2 | NP_060352.1 | 4 | 494 | ||
| GenBank mRNA | AK000568.1 | - | 4 | 439 | ||
| AK027604.1 | - | 4 | 462 | |||
| AK291175.1 | - | 4 | 421 | |||
| AK293197.1 | - | 4 | 670 | |||
| BC010849.1 | - | 4 | 408 | 3878776 | ||
| BC013130.1 | - | 4 | 345 | 4517748 | ||
| CR457244.1 | - | 4 | 336 | |||
| KU178671.1 | - | 4 | 336 | |||
| KU178672.1 | - | 4 | 336 | |||
| KU178673.1 | - | 3 | 237 | |||

CLN6
Genomic Map