siRNA Details
| Gene Symbol: C1orf123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304759.1 | NP_001291688.1 | 3 | 194 | ||
| NM_001304760.1 | NP_001291689.1 | 2 | 110 | |||
| NM_017887.2 | NP_060357.1 | 4 | 251 | |||
| GenBank mRNA | AK000587.1 | - | 4 | 229 | ||
| BC010908.1 | - | 4 | 233 | 4093763 | ||
| BE889129.1 | - | 1 | 63 | |||
| BI603716.1 | - | 3 | 187 | |||
| CR457246.1 | - | 3 | 187 | |||

C1orf123
Genomic Map