siRNA Details
Gene Symbol: ZNF770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014106.3 | NP_054825.2 | 3 | 2024 | ||
XM_011521744.2 | XP_011520046.1 | 2 | 1868 | |||
GenBank mRNA | AK127201.1 | - | 1 | 135 | ||
AK131433.1 | - | 1 | 896 | |||
AK291438.1 | - | 3 | 2015 | |||
BC071603.1 | - | 3 | 1996 | 30340520 | ||
BM454360.1 | - | 1 | 545 |
