siRNA Details
| Gene Symbol: TESC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168325.1 | NP_001161797.1 | 1, 2 | 227 | ||
| NM_017899.3 | NP_060369.3 | 1, 2 | 227 | |||
| NR_031766.2 | 1, 2 | 227 | ||||
| XM_017019532.1 | XP_016875021.1 | 1, 2 | 271 | |||
| XM_017019533.1 | XP_016875022.1 | 1, 2 | 271 | |||
| GenBank mRNA | AF443207.1 | - | 1, 2 | 184 | ||
| AK000614.1 | - | 1, 2 | 270 | |||
| BC015221.1 | - | 1, 2 | 184 | 3885280 | ||
| BQ434057.1 | - | 1, 2 | 160 | |||
| BQ949772.1 | - | 1, 2 | 181 | |||
| DB452909.1 | - | 1, 2 | 227 | |||

TESC
Genomic Map