siRNA Details
| Gene Symbol: LAMTOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017907.2 | NP_060377.1 | 5 | 1092 | ||
| GenBank mRNA | AF289583.1 | - | 5 | 983 | ||
| AK000632.1 | - | 5 | 981 | |||
| BC001706.1 | - | 5 | 954 | 3544000 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LAMTOR1| Gene Symbol: LAMTOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017907.2 | NP_060377.1 | 5 | 1092 | ||
| GenBank mRNA | AF289583.1 | - | 5 | 983 | ||
| AK000632.1 | - | 5 | 981 | |||
| BC001706.1 | - | 5 | 954 | 3544000 | ||