siRNA Details
| Gene Symbol: C14orf119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017924.3 | NP_060394.1 | 2 | 1529 | ||
| GenBank mRNA | BC009645.1 | - | 2 | 1424 | 3458006 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C14orf119| Gene Symbol: C14orf119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017924.3 | NP_060394.1 | 2 | 1529 | ||
| GenBank mRNA | BC009645.1 | - | 2 | 1424 | 3458006 | |