siRNA Details
| Gene Symbol: TTC38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017931.2 | NP_060401.2 | 7, 8 | 815 | ||
| XM_011530259.1 | XP_011528561.1 | 8, 9 | 943 | |||
| GenBank mRNA | AK000706.1 | - | 7, 8 | 771 | ||
| AK300606.1 | - | 6, 7 | 621 | |||
| BC018918.2 | - | 7, 8 | 756 | 4130906 | ||

TTC38
Genomic Map