siRNA Details
| Gene Symbol: CHI3L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025197.1 | NP_001020368.1 | 8, 9 | 1080 | ||
| NM_001025199.1 | NP_001020370.1 | 8, 9 | 1140 | |||
| NM_004000.2 | NP_003991.2 | 9, 10 | 1110 | |||
| GenBank mRNA | AK223316.1 | - | 8, 9 | 1020 | ||
| AK298466.1 | - | 8, 9 | 1031 | |||
| BC011460.1 | - | 9, 10 | 1048 | 4214121 | ||
| BT006767.1 | - | 9, 10 | 1039 | |||
| U49835.1 | - | 9, 10 | 1059 | |||
| U58514.1 | - | 9, 10 | 1075 | |||
| U58515.1 | - | 8, 9 | 1141 | |||

CHI3L2
Genomic Map