siRNA Details
| Gene Symbol: HEATR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182922.2 | NP_891552.1 | 9 | 1348 | ||
| XM_005256013.2 | XP_005256070.1 | 8 | 1163 | |||
| XM_011523188.2 | XP_011521490.1 | 9 | 1238 | |||
| XM_017023385.1 | XP_016878874.1 | 7 | 1301 | |||
| XM_017023386.1 | XP_016878875.1 | 7 | 1106 | |||
| GenBank mRNA | AF462442.1 | - | 7 | 1261 | ||
| AK000725.1 | - | 8 | 1155 | |||
| AK289949.1 | - | 9 | 1313 | |||
| BC014415.1 | - | 1325 | ||||
| BC018730.1 | - | 9 | 1313 | 4907047 | ||
| BC033077.1 | - | 9 | 1214 | 4301559 | ||

HEATR3
Genomic Map