siRNA Details
| Gene Symbol: USP47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282659.1 | NP_001269588.1 | 15, 16 | 2520 | ||
| NM_017944.3 | NP_060414.3 | 14, 15 | 2316 | |||
| XM_005252997.1 | XP_005253054.1 | 16, 17 | 2580 | |||
| XM_011520196.1 | XP_011518498.1 | 8, 9 | 1695 | |||
| XM_017017953.1 | XP_016873442.1 | 16, 17 | 2636 | |||
| XM_017017954.1 | XP_016873443.1 | 8, 9 | 2843 | |||
| GenBank mRNA | AK027362.1 | - | 14, 15 | 1721 | ||
| AK092290.1 | - | 16, 17 | 1936 | |||
| AK127264.1 | - | 15, 16 | 2316 | |||
| BG752249.1 | - | 350 | ||||

USP47
Genomic Map