siRNA Details
| Gene Symbol: VPS37C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017966.4 | NP_060436.4 | 3 | 258 | ||
| XM_005274077.3 | XP_005274134.1 | 3 | 299 | |||
| GenBank mRNA | AK097326.1 | - | 3 | 235 | ||
| AK302384.1 | - | 3 | 326 | |||
| AL834261.1 | - | 3 | 266 | |||
| BC141827.1 | - | 3 | 221 | 40148285 | ||
| BC142702.1 | - | 3 | 221 | 40148798 | ||
| BM450171.1 | - | 2 | 228 | |||

VPS37C
Genomic Map