siRNA Details
| Gene Symbol: NRDE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017970.3 | NP_060440.2 | 9, 10 | 2083 | ||
| XM_011536886.1 | XP_011535188.1 | 10, 11 | 1911 | |||
| XM_017021394.1 | XP_016876883.1 | 10, 11 | 1911 | |||
| XM_017021395.1 | XP_016876884.1 | 10, 11 | 2032 | |||
| XM_017021396.1 | XP_016876885.1 | 9, 10 | 1626 | |||
| GenBank mRNA | AK024113.1 | - | 4, 5 | 624 | ||
| AK294958.1 | - | 9, 10 | 1887 | |||
| BC098568.1 | - | 9, 10 | 1861 | 30706812 | ||
| BC118673.1 | - | 6, 7 | 1416 | 40028659 | ||

NRDE2
Genomic Map