siRNA Details
| Gene Symbol: MRPL20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318485.1 | NP_001305414.1 | 1 | 154 | ||
| NM_017971.3 | NP_060441.2 | 1 | 154 | |||
| GenBank mRNA | AB049644.1 | - | 1 | 85 | ||
| AK000886.1 | - | 1 | 88 | |||
| AK301440.1 | - | 1 | 134 | |||
| AK315794.1 | - | 1 | 88 | |||
| BC009515.1 | - | 1 | 122 | 3542715 | ||
| BC014316.1 | - | 1 | 91 | 4045076 | ||
| BC059945.1 | - | 1 | 72 | 5397911 | ||
| BG700526.1 | - | 1 | 155 | |||
| DN990876.1 | - | 1 | 126 | |||

MRPL20
Genomic Map