siRNA Details
| Gene Symbol: SUSD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037175.2 | NP_001032252.1 | 4 | 555 | ||
| NM_017982.3 | NP_060452.3 | 4 | 555 | |||
| XM_005273169.1 | XP_005273226.1 | 5 | 572 | |||
| XM_005273172.1 | XP_005273229.1 | 5 | 635 | |||
| XM_006711408.1 | XP_006711471.1 | 4 | 553 | |||
| XM_011509685.1 | XP_011507987.1 | 4 | 587 | |||
| XM_011509687.1 | XP_011507989.1 | 5 | 635 | |||
| XM_017001583.1 | XP_016857072.1 | 5 | 753 | |||
| XM_017001584.1 | XP_016857073.1 | 5 | 752 | |||
| XM_017001585.1 | XP_016857074.1 | 6 | 736 | |||
| XM_017001586.1 | XP_016857075.1 | 5 | 630 | |||
| XM_017001587.1 | XP_016857076.1 | 5 | 572 | |||
| XM_017001588.1 | XP_016857077.1 | 4 | 587 | |||
| XM_017001589.1 | XP_016857078.1 | 5 | 757 | |||
| GenBank mRNA | AK000914.1 | - | 4 | 436 | ||
| AK096265.1 | - | 3 | 1009 | |||
| AK295553.1 | - | 3 | 305 | |||
| AK296923.1 | - | 5 | 635 | |||
| AY358495.1 | - | 4 | 532 | |||
| BC004888.2 | - | 4 | 436 | 3926397 | ||
| EF560748.1 | - | 4 | 483 | |||

SUSD4
Genomic Map