siRNA Details
| Gene Symbol: TMEM248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017994.4 | NP_060464.1 | 5, 6 | 1053 | ||
| XM_005250482.3 | XP_005250539.1 | 5, 6 | 905 | |||
| GenBank mRNA | AK000961.1 | - | 5, 6 | 1039 | ||
| AK222774.1 | - | 5, 6 | 1053 | |||
| BC008675.1 | - | 5, 6 | 1033 | 3853834 | ||
| BC012562.1 | - | 5, 6 | 1048 | 4332094 | ||
| BC041764.2 | - | 5, 6 | 1022 | 4152659 | ||

TMEM248
Genomic Map