siRNA Details
| Gene Symbol: TMEM45A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018004.2 | NP_060474.1 | 4 | 834 | ||
| XM_005247569.2 | XP_005247626.1 | 5 | 795 | |||
| XM_017006715.1 | XP_016862204.1 | 5 | 955 | |||
| GenBank mRNA | AB013911.1 | - | 3 | 521 | ||
| AK000996.1 | - | 4 | 834 | |||
| AK129789.1 | - | 4 | 684 | |||
| AY450392.1 | - | 4 | 834 | |||
| BC040355.1 | - | 5 | 768 | 4748104 | ||

TMEM45A
Genomic Map