siRNA Details
| Gene Symbol: IFT57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018010.3 | NP_060480.1 | 2 | 497 | ||
| GenBank mRNA | AB014762.1 | - | 2 | 249 | ||
| AF139576.1 | - | 2 | 327 | |||
| AF245220.1 | - | 2 | 391 | |||
| AK001009.1 | - | 2 | 336 | |||
| AK298854.1 | - | 2 | 395 | |||
| BC011899.2 | - | 2 | 346 | 4053749 | ||
| CR457251.1 | - | 2 | 249 | |||
| DA362330.1 | - | 2 | 497 | |||

IFT57
Genomic Map