siRNA Details
| Gene Symbol: CXorf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018015.5 | NP_060485.4 | 8 | 1710 | ||
| GenBank mRNA | AK001040.1 | - | 8 | 1665 | ||
| AK291730.1 | - | 8 | 1659 | |||
| BC011483.1 | - | 8 | 1631 | 3453700 | ||
| BC033709.1 | - | 8 | 1622 | 3910777 | ||
| BC070110.1 | - | 8 | 1661 | 5264812 | ||

CXorf57
Genomic Map