siRNA Details
| Gene Symbol: SAMD4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303614.1 | NP_001290543.1 | 8 | 1799 | ||
| NM_018028.3 | NP_060498.2 | 11 | 2520 | |||
| XM_011527057.2 | XP_011525359.1 | 9 | 1810 | |||
| XM_011527060.2 | XP_011525362.1 | 9 | 2912 | |||
| XM_011527061.2 | XP_011525363.1 | 8 | 1724 | |||
| XM_011527062.2 | XP_011525364.1 | 8 | 1785 | |||
| XM_011527063.2 | XP_011525365.1 | 9 | 1809 | |||
| XM_011527064.2 | XP_011525366.1 | 9 | 1810 | |||
| XM_017026918.1 | XP_016882407.1 | 8 | 1744 | |||
| XM_017026919.1 | XP_016882408.1 | 10 | 1900 | |||
| XM_017026920.1 | XP_016882409.1 | 10 | 1922 | |||
| XM_017026921.1 | XP_016882410.1 | 8 | 1785 | |||
| XM_017026922.1 | XP_016882411.1 | 10 | 1902 | |||
| XM_017026923.1 | XP_016882412.1 | 10 | 7440 | |||
| GenBank mRNA | BC039652.1 | - | 4 | 497 | ||
| BC054518.1 | - | 11 | 2511 | 6501489 | ||
| BC065211.1 | - | 5 | 807 | |||
| BC080641.1 | - | 8 | 1759 | 6645022 | ||
| EF601121.1 | - | 11 | 2507 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_175021, BC042901.

SAMD4B
Genomic Map