siRNA Details
| Gene Symbol: SLFN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289009.1 | NP_001275938.1 | 4 | 1734 | ||
| NM_018042.4 | NP_060512.3 | 4 | 1799 | |||
| XM_005257995.4 | XP_005258052.1 | 5 | 1690 | |||
| XM_011524967.2 | XP_011523269.1 | 6 | 1812 | |||
| XM_017024809.1 | XP_016880298.1 | 4 | 1623 | |||
| XM_017024810.1 | XP_016880299.1 | 5 | 2280 | |||
| XM_017024811.1 | XP_016880300.1 | 7 | 1931 | |||
| GenBank mRNA | AK001122.1 | - | 4 | 1775 | ||
| AK291826.1 | - | 4 | 1712 | |||
| BC035605.1 | - | 4 | 1799 | 5163323 | ||
| BX647081.1 | - | 7 | 1922 | |||

SLFN12
Genomic Map