siRNA Details
| Gene Symbol: AGGF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018046.4 | NP_060516.2 | 4 | 1002 | ||
| GenBank mRNA | AW958325.1 | - | 4 | 484 | ||
| AY500994.1 | - | 4 | 921 | |||
| BC032844.1 | - | 4 | 988 | 5266978 | ||
| BC073864.1 | - | 4 | 529 | |||
| BE077189.1 | - | 2 | 277 | |||

AGGF1
Genomic Map