siRNA Details
| Gene Symbol: CRTAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206528.2 | NP_001193457.1 | 3 | 597 | ||
| NM_018058.6 | NP_060528.3 | 3 | 597 | |||
| XM_005269938.4 | XP_005269995.1 | 3 | 390 | |||
| XM_011539917.1 | XP_011538219.1 | 3 | 390 | |||
| XM_017016366.1 | XP_016871855.1 | 3 | 390 | |||
| XM_017016367.1 | XP_016871856.1 | 3 | 390 | |||
| GenBank mRNA | AJ276171.1 | - | 3 | 390 | ||
| AJ279016.1 | - | 2 | 217 | |||
| AJ421515.1 | - | 2 | 217 | |||
| AK057190.1 | - | 3 | 597 | |||
| AK292084.1 | - | 3 | 359 | |||

CRTAC1
Genomic Map