siRNA Details
| Gene Symbol: HEATR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018072.5 | NP_060542.4 | 13 | 1754 | ||
| GenBank mRNA | AK095692.1 | - | 13 | 1717 | ||
| BC150614.1 | - | 12 | 1624 | 9056984 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HEATR1| Gene Symbol: HEATR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018072.5 | NP_060542.4 | 13 | 1754 | ||
| GenBank mRNA | AK095692.1 | - | 13 | 1717 | ||
| BC150614.1 | - | 12 | 1624 | 9056984 | ||