siRNA Details
| Gene Symbol: LRRC8D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134479.1 | NP_001127951.1 | 3 | 2117 | ||
| NM_018103.4 | NP_060573.2 | 3 | 2104 | |||
| XM_011541689.1 | XP_011539991.1 | 4 | 2188 | |||
| XM_017001599.1 | XP_016857088.1 | 3 | 2459 | |||
| XM_017001600.1 | XP_016857089.1 | 3 | 1914 | |||
| XM_017001601.1 | XP_016857090.1 | 3 | 2805 | |||
| XM_017001602.1 | XP_016857091.1 | 3 | 2158 | |||
| GenBank mRNA | AK001332.1 | - | 1 | 1170 | ||
| AK092238.1 | - | 3 | 2080 | |||
| AK222787.1 | - | 1 | 1131 | |||
| AL831880.1 | - | 1 | 1576 | |||
| AL832952.1 | - | 1 | 1535 | |||
| AY358874.1 | - | 1 | 692 | |||
| BC024159.2 | - | 3 | 2071 | 3958069 | ||

LRRC8D
Genomic Map