siRNA Details
Gene Symbol: DALRD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009996.2 | NP_001009996.1 | 5 | 945 | ||
NM_001276405.1 | NP_001263334.1 | 5 | 945 | |||
NM_018114.5 | NP_060584.3 | 5 | 962 | |||
XM_005265269.3 | XP_005265326.1 | 5 | 926 | |||
XM_006713219.2 | XP_006713282.1 | 6 | 1081 | |||
XM_011533891.1 | XP_011532193.1 | 5 | 926 | |||
XM_011533892.1 | XP_011532194.1 | 5 | 1277 | |||
XM_011533893.1 | XP_011532195.1 | 5 | 926 | |||
XM_017006723.1 | XP_016862212.1 | 6 | 1081 | |||
GenBank mRNA | BC047683.1 | - | 5 | 907 | 5755743 |
