siRNA Details
| Gene Symbol: KCNC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039574.2 | NP_001034663.1 | 2 | 1711 | ||
| NM_004978.4 | NP_004969.2 | 2 | 1711 | |||
| NR_036437.1 | 2 | 1711 | ||||
| XM_006710625.3 | XP_006710688.1 | 2 | 1555 | |||
| XM_006710627.3 | XP_006710690.1 | 2 | 1555 | |||
| XM_011541401.2 | XP_011539703.1 | 2 | 1555 | |||
| GenBank mRNA | AK310193.1 | - | 2 | 1136 | ||
| BC101769.1 | - | 2 | 982 | 8069275 | ||
| BX537638.1 | - | 1 | 3499 | |||
| M64676.1 | - | 2 | 1081 | |||
Citations (1)
p21-activated kinase (Pak) regulates NADPH oxidase activation in human neutrophils.
- Authors
- Martyn KD, Kim MJ, Quinn MT, Dinauer MC, Knaus UG
- Journal
- Blood - Page No 3962 (Thu Dec 01 00:00:00 UTC 2005)
- Volume (Year)
- 106 (2005)
- Product Usage
- -

KCNC4
Genomic Map