siRNA Details
| Gene Symbol: LYST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000081.3 | NP_000072.2 | 18 | 5684 | ||
| NM_001301365.1 | NP_001288294.1 | 18 | 6121 | |||
| XM_011544031.1 | XP_011542333.1 | 18 | 5684 | |||
| XM_011544032.1 | XP_011542334.1 | 18 | 6087 | |||
| XM_011544033.2 | XP_011542335.1 | 18 | 5834 | |||
| XM_011544034.1 | XP_011542336.1 | 18 | 5684 | |||
| XM_011544035.2 | XP_011542337.1 | 18 | 5684 | |||
| XM_011544036.2 | XP_011542338.1 | 14 | 3338 | |||
| XM_011544037.2 | XP_011542339.1 | 18 | 5684 | |||
| XM_011544039.2 | XP_011542341.1 | 18 | 5684 | |||
| XM_011544040.2 | XP_011542342.1 | 18 | 5684 | |||
| XM_017000150.1 | XP_016855639.1 | 18 | 5684 | |||
| XM_017000151.1 | XP_016855640.1 | 18 | 5684 | |||
| GenBank mRNA | BC144702.1 | - | 17 | 5558 | ||
| U67615.1 | - | 18 | 5691 | |||
| U70064.1 | - | 18 | 6054 | |||

LYST
Genomic Map