siRNA Details
| Gene Symbol: MSTO2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024117.2 | 4 | 399 | |||
| GenBank mRNA | AK293928.1 | - | 4 | 399 | ||
| AK295146.1 | - | 4 | 399 | |||
| BC099909.1 | - | 4 | 349 | |||
| DC387604.1 | - | 4 | 399 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Gene Symbol: MSTO2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024117.2 | 4 | 399 | |||
| GenBank mRNA | AK293928.1 | - | 4 | 399 | ||
| AK295146.1 | - | 4 | 399 | |||
| BC099909.1 | - | 4 | 349 | |||
| DC387604.1 | - | 4 | 399 | |||