siRNA Details
| Gene Symbol: TMEM33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018126.2 | NP_060596.2 | 3 | 596 | ||
| XM_005248116.3 | XP_005248173.1 | 4 | 435 | |||
| XM_005248117.2 | XP_005248174.1 | 4 | 383 | |||
| GenBank mRNA | AK001387.1 | - | 3 | 564 | ||
| AK023062.1 | - | 3 | 576 | |||
| AK094190.1 | - | 4 | 507 | |||
| AY659966.1 | - | 3 | 564 | |||
| BC000948.2 | - | 3 | 326 | 3448888 | ||
| CR457260.1 | - | 3 | 231 | |||

TMEM33
Genomic Map