siRNA Details
| Gene Symbol: CEP55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127182.1 | NP_001120654.1 | 6 | 1072 | ||
| NM_018131.4 | NP_060601.3 | 6 | 1194 | |||
| XM_011539918.1 | XP_011538220.1 | 6 | 1208 | |||
| XM_011539919.1 | XP_011538221.1 | 6 | 1076 | |||
| XM_011539920.2 | XP_011538222.1 | 6 | 902 | |||
| XM_017016372.1 | XP_016871861.1 | 6 | 1048 | |||
| XM_017016373.1 | XP_016871862.1 | 6 | 1070 | |||
| GenBank mRNA | AB091343.1 | - | 6 | 1183 | ||
| AK001402.1 | - | 5 | 794 | |||
| AK223495.1 | - | 6 | 1175 | |||
| AK315536.1 | - | 6 | 1191 | |||
| AY788918.1 | - | 6 | 1194 | |||
| BC008947.2 | - | 6 | 1174 | 3028566 | ||

CEP55
Genomic Map