siRNA Details
| Gene Symbol: CHRM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320917.1 | NP_001307846.1 | 2 | 2212 | ||
| NM_012125.3 | NP_036257.1 | 3 | 1970 | |||
| GenBank mRNA | AB084282.1 | - | 2 | 1619 | ||
| AF026263.1 | - | 1 | 1300 | |||
| AF385591.1 | - | 1 | 1300 | |||
| AF498919.1 | - | 1 | 1300 | |||
| AK095198.1 | - | 3 | 1970 | |||
| BC041805.1 | - | 2 | 1614 | 5298446 | ||
| BC068528.1 | - | 2 | 1608 | 30339053 | ||
More Information
Set Membership: |
|

CHRM5
Genomic Map