siRNA Details
| Gene Symbol: INTS10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018142.2 | NP_060612.2 | 6 | 691 | ||
| XM_005273555.3 | XP_005273612.1 | 6 | 949 | |||
| XM_005273556.3 | XP_005273613.1 | 6 | 733 | |||
| XM_005273557.3 | XP_005273614.1 | 6 | 733 | |||
| XM_006716355.2 | XP_006716418.1 | 6 | 838 | |||
| XM_017013606.1 | XP_016869095.1 | 6 | 733 | |||
| XM_017013607.1 | XP_016869096.1 | 6 | 949 | |||
| XM_017013608.1 | XP_016869097.1 | 6 | 514 | |||
| XM_017013609.1 | XP_016869098.1 | 6 | 838 | |||
| GenBank mRNA | AK001431.1 | - | 6 | 611 | ||
| AK092931.1 | - | 2 | 1609 | |||
| BC001915.1 | - | 4 | 328 | |||
| BC006209.2 | - | 4 | 328 | |||
| BC051282.1 | - | 4 | 342 | |||
| BM547763.1 | - | 6 | 692 | |||
| CR457262.1 | - | 3 | 221 | |||

INTS10
Genomic Map