siRNA Details
| Gene Symbol: SEC61A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142627.2 | NP_001136099.1 | 8 | 873 | ||
| NM_001142628.1 | NP_001136100.1 | 7 | 807 | |||
| NM_018144.3 | NP_060614.2 | 8 | 873 | |||
| NR_024576.2 | 8 | 873 | ||||
| NR_024577.2 | 9 | 1006 | ||||
| GenBank mRNA | AF346603.1 | - | 8 | 814 | ||
| AK001440.1 | - | 1 | 108 | |||
| AK022027.1 | - | 2 | 256 | |||
| AK022640.1 | - | 8 | 808 | |||
| AK057532.1 | - | 9 | 947 | |||
| AK292295.1 | - | 8 | 823 | |||
| AK301841.1 | - | 7 | 760 | |||
| BC026179.1 | - | 8 | 823 | 4823130 | ||

SEC61A2
Genomic Map