siRNA Details
| Gene Symbol: RMDN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304802.1 | NP_001291731.1 | 12 | 1545 | ||
| NM_001323894.1 | NP_001310823.1 | 12 | 1459 | |||
| NM_001323895.1 | NP_001310824.1 | 12 | 1676 | |||
| NM_001323896.1 | NP_001310825.1 | 12 | 1537 | |||
| NM_001323897.1 | NP_001310826.1 | 12 | 1653 | |||
| NM_018145.2 | NP_060615.1 | 12 | 1575 | |||
| XM_011521755.2 | XP_011520057.1 | 9 | 1821 | |||
| GenBank mRNA | AB000782.1 | - | 11 | 1314 | ||
| AK001441.1 | - | 12 | 1499 | |||
| AK025963.1 | - | 9 | 888 | |||
| AK090731.1 | - | 5 | 1447 | |||
| AK092058.1 | - | 12 | 1458 | |||
| AK097286.1 | - | 13 | 1387 | |||
| AK123192.1 | - | 13 | 1452 | |||
| AK123282.1 | - | 11 | 2382 | |||
| AY358793.1 | - | 11 | 1315 | |||
| BC008970.2 | - | 8 | 772 | |||
| BC063844.1 | - | 12 | 1463 | 6527521 | ||

RMDN3
Genomic Map