siRNA Details
| Gene Symbol: RMDN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304802.1 | NP_001291731.1 | 2 | 392 | ||
| NM_001323894.1 | NP_001310823.1 | 2 | 306 | |||
| NM_001323895.1 | NP_001310824.1 | 2 | 422 | |||
| NM_001323896.1 | NP_001310825.1 | 2 | 306 | |||
| NM_001323897.1 | NP_001310826.1 | 2 | 422 | |||
| NM_018145.2 | NP_060615.1 | 2 | 422 | |||
| GenBank mRNA | AB000782.1 | - | 1 | 161 | ||
| AK001441.1 | - | 2 | 346 | |||
| AK092058.1 | - | 2 | 305 | |||
| AK123192.1 | - | 2 | 368 | |||
| AK123282.1 | - | 1 | 1229 | |||
| AY358793.1 | - | 1 | 162 | |||
| BC063844.1 | - | 2 | 310 | 6527521 | ||
| DA280017.1 | - | 2 | 278 | |||
| DA592879.1 | - | 2 | 306 | |||

RMDN3
Genomic Map