siRNA Details
| Gene Symbol: SMG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018149.6 | NP_060619.4 | 3 | 2740 | ||
| GenBank mRNA | AF349467.1 | - | 3 | 2740 | ||
| AK001449.1 | - | 3 | 1889 | |||
| AK026858.1 | - | 3 | 2713 | |||
| AL834490.1 | - | 3 | 2719 | |||
| BC020957.1 | - | 3 | 1908 | 3851898 | ||

SMG8
Genomic Map