siRNA Details
| Gene Symbol: VPS13D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015378.3 | NP_056193.2 | 19 | 3156 | ||
| NM_018156.3 | NP_060626.2 | 19 | 3156 | |||
| GenBank mRNA | AJ608774.1 | - | 19 | 3128 | ||
| AJ608775.1 | - | 19 | 3128 | |||
| AL162331.1 | - | 19 | 3128 | |||

VPS13D
Genomic Map